missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Base Oil 20, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
500 mL
1 gal.
Unit Size:
3.80 L
500mL
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As(+5) | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Chemical Name or Material | Base Oil 20 Blank |
Packaging | Triple-leached LDPE bottle |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Type | Certified Reference Material |
Solvent or Matrix | Base Oil 20 |
Industry Type | Petrochemical Testing |
Quantity | 1 gal. |
Safety and Handling

Product Identifier
- Base Oil 20 Standard
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : white mineral oil
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Base Oil 20, SPEX CertiPrep™ >
Spot an opportunity for improvement?Share a Content Correction