Learn More
Heidolph™ Clamp for Erlenmeyer Flasks
Clamps for use with various perforated platforms
Brand:  Heidolph™ 549-63000-00
 View more versions of this product
Product Code. 11711659
  • £70.40 / Each
Only null left
Estimated Shipment: 28-07-2025
Add to basket
Description
Description
  • Maximum number of clamps per platform type:
    • 549-51000-00
      • For platform 100 - max. 16
      • For platform 1000 - max. 20
      • For platform 2000 - max. 36
    • 549-52000-00
      • For platform 100 - max. 16
      • For platform 1000 - max. 20
      • For platform 2000 - max. 36
    • 549-55000-00
      • For platform 100 - max. 3
      • For platform 1000 - max. 4
      • For platform 2000 - max. 9
    • 549-56000-00
      • For platform 100 - max. 2
      • For platform 1000 - max. 4
      • For platform 2000 - max. 5
    • 549-63000-00
      • For platform 1000 - max. 2
      • For platform 2000 - max. 3
      TRUSTED_SUSTAINABILITY
Specifications
Specifications
2000 mL Erlenmeyer flasks
Clamp
Perforated platforms 1000 and 2000
7