missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Brand:  Thermo Scientific™ 150472

 View more versions of this product

Product Code. 30408804

  • £178.00 / Pack of 280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Clear
Single-use
Polystyrene
Sterile
280 Dishes
16 mm
Flat
21.5 cm2
Not Gridded
Round
Cell Culture Dish
10 Dishes
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use