Learn More
SPAG5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14726-1-AP-20UL
 View more versions of this product
Product Code. 16883514
  • £135.00 / 20µL
Description
This antibody is specific to SPAG5.
SPAG5 (sperm associated antigen 5), also known as MAP126, hMAP126, DEEPEST or Astrin, is a 1,193 amino acid protein that is expressed at high levels in testis and at lower levels in liver, placenta, pancreas, colon and thymus. Localized specifically to spindle poles during prophase and throughout the spindle during metaphase and anaphase, SPAG5 is essential for proper spindle formation and is thought to be involved in the dynamic and functional regulation of mitotic spindles. Additionally, SPAG5 plays a role in sister chromatid cohesion and subsequent chromatid separation and, via its association with chromatids, is an important regulator of chromosome integrity. SPAG5 has a long stalk and a globular head domain through which if forms a homodimer that is characterized by aster-like structures. Upon DNA damage, SPAG5 may be phosphorylated by ATR or ATM.
Specifications
Specifications
SPAG5
Polyclonal
Unconjugated
SPAG5
Astrin, DEEPEST, hMAP126, MAP126, SPAG5, sperm associated antigen 5
Rabbit
Antigen Affinity Chromatography
RUO
10615
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96R06
SPAG5
SPAG5 Fusion Protein Ag6485
20 μL