Learn More
BINDER™ Standard Rack
Shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
Brand: BINDER™ 8012-2389
This item is not returnable.
View return policy
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Specifications
KB 130, KB PRO 130, KBF 130, KBF PRO 130 | |
Stainless Steel | |
Rack |